3-Methylenecyclobutanecarbonitrile ≥95%

Supplier: Matrix Scientific

059132-5G 059132-25G
101917-216EA 96.92 USD
101917-216 101917-218
3-Methylenecyclobutanecarbonitrile ≥95%
3-Methylenecyclobutanecarbonitrile

Hazard: Irritant

Formula: C₆H₇N
MW: 93.13 g/mol
MDL Number: MFCD01085401
CAS Number: 15760-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR