Methyl acetopyruvate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2,4-dioxopentanoate

059131-5G 059131-25G
101917-210EA 73.85 USD
101917-210 101917-212
Methyl acetopyruvate ≥95%
Methyl acetopyruvate

Hazard: Irritant

Formula: C₆H₈O₄
MW: 144.13 g/mol
MDL Number: MFCD00043638
CAS Number: 20577-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR