Methyl (4,6-dihydroxypyrimidin-2-ylsulfanyl)acetate ≥95%

Supplier: Matrix Scientific

059130-1G 059130-5G
101917-206EA 349.23 USD
101917-206 101917-208
Methyl (4,6-dihydroxypyrimidin-2-ylsulfanyl)acetate ≥95%
Methyl (4,6-dihydroxypyrimidin-2-ylsulfanyl)acetate

Hazard: Irritant

Formula: C₇H₈N₂O₄S
MW: 216.22 g/mol
MDL Number: MFCD00489772
CAS Number: 370077-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR