Methyl (3-chloro-5-(trifluoromethyl)pyridin-2-ylsulfanyl)acetate ≥95%
Supplier: Matrix Scientific
059129-1G
059129-5G
101917-202EA
436.92
USD
101917-202
101917-204
Methyl (3-chloro-5-(trifluoromethyl)pyridin-2-ylsulfanyl)acetate ≥95%
Methyl (3-chloro-5-(trifluoromethyl)pyridin-2-ylsulfanyl)acetate
Hazard: Irritant
Formula:
C₉H₇ClF₃NO₂S |
MDL Number:
MFCD00245199 CAS Number: 1024368-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...