Methyl (6-chloro-4-(trifluoromethyl)pyridin-2-ylsulfanyl)acetate ≥95%
Supplier: Matrix Scientific
059127-1G
059127-5G
101917-194EA
553.85
USD
101917-194
101917-196
Methyl (6-chloro-4-(trifluoromethyl)pyridin-2-ylsulfanyl)acetate ≥95%
Methyl (6-chloro-4-(trifluoromethyl)pyridin-2-ylsulfanyl)acetate
Hazard: Irritant
Formula:
C₉H₇ClF₃NO₂S |
MDL Number:
MFCD11052385 CAS Number: 1053656-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...