1-(4-Methoxypyridin-2-yl)ethanone ≥95%

Supplier: Matrix Scientific

059122-1G 059122-5G
101917-178EA 593.85 USD
101917-178 101917-180
1-(4-Methoxypyridin-2-yl)ethanone ≥95%
1-(4-Methoxypyridin-2-yl)ethanone

Hazard: Irritant

Formula: C₈H₉NO₂
MDL Number: MFCD10697664
CAS Number: 59576-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR