2-(3-Methoxyphenyl)thioacetamide ≥95%
Supplier: Matrix Scientific
059120-1G
059120-5G
101917-170EA
349.23
USD
101917-170
101917-172
2-(3-Methoxyphenyl)thioacetamide ≥95%
2-(3-Methoxyphenyl)thioacetamide
Hazard: Irritant
Formula:
C₉H₁₁NOS |
MDL Number:
MFCD09414738 CAS Number: 35582-11-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...