(4-Methoxyphenyl)morpholin-4-ylacetonitrile hydrochloride ≥95%

Supplier: Matrix Scientific

059119-1G 059119-5G 059119-10G
101917-164EA 204.62 USD
101917-164 101917-166 101917-168
(4-Methoxyphenyl)morpholin-4-ylacetonitrile hydrochloride ≥95%
(4-Methoxyphenyl)morpholin-4-ylacetonitrile hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₇ClN₂O₂
MDL Number: MFCD11052435
CAS Number: 1129307-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR