Methyl-6-methoxy-1H-indole-2-carboxylate ≥95%

Supplier: Matrix Scientific

059116-1G 059116-5G
101917-150EA 36.92 USD
101917-150 101917-152
Methyl-6-methoxy-1H-indole-2-carboxylate ≥95%
Methyl-6-methoxy-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₁NO₃
MDL Number: MFCD00134301
CAS Number: 98081-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR