N'-[1-(2-Methoxyethylamino)-2-phenoxyethylidene]hydrazine carboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

059115-500MG 059115-1G
101917-146EA 284.62 USD
101917-146 101917-148
N'-[1-(2-Methoxyethylamino)-2-phenoxyethylidene]hydrazine carboxylic acid-tert-butyl ester ≥95%
N'-[1-(2-Methoxyethylamino)-2-phenoxyethylidene]hydrazine carboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₆H₂₅N₃O₄
MW: 323.4 g/mol
MDL Number: MFCD10568262
CAS Number: 1053658-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR