2-Methoxybenzamidine hydrochloride ≥95%
Supplier: Matrix Scientific
059113-1G
059113-5G
101917-136EA
284.62
USD
101917-136
101917-138
2-Methoxybenzamidine hydrochloride ≥95%
2-Methoxybenzamidine hydrochloride
Hazard: Irritant
Formula:
C₈H₁₁ClN₂O MW: 186.64 g/mol |
MDL Number:
MFCD06738644 CAS Number: 51818-19-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...