3-Methoxybenzaldehyde oxime ≥95%

Supplier: Matrix Scientific

059112-1G 059112-5G
101917-132EA 295.38 USD
101917-132 101917-134
3-Methoxybenzaldehyde oxime ≥95%
3-Methoxybenzaldehyde oxime

Hazard: Irritant

Formula: C₈H₉NO₂
MDL Number: MFCD02256033
CAS Number: 38489-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR