Isoxazol-5-ylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Aminoisoxazole, Isoxazol-5-amine

059110-500MG 059110-1G
101917-126EA 153.85 USD
101917-126 101917-128
Isoxazol-5-ylamine ≥95%
Isoxazol-5-ylamine

Hazard: Irritant

Formula: C₃H₄N₂O
MW: 84.08 g/mol
MDL Number: MFCD00128192
CAS Number: 14678-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR