1-Indan-2-ylethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Indan-2-yl-ethanone

059106-1G 059106-5G
101917-112EA 683.08 USD
101917-112 101917-114
1-Indan-2-ylethanone ≥95%
1-Indan-2-ylethanone

Hazard: Irritant

Formula: C₁₁H₁₂O
MW: 160.22 g/mol
MDL Number: MFCD08543446
CAS Number: 33982-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR