3-Imidazol-1-ylphenylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(1H-Imidazol-1-yl)aniline

059105-1G 059105-5G 059105-10G
101917-106EA 247.69 USD
101917-106 101917-108 101917-110
3-Imidazol-1-ylphenylamine ≥95%
3-Imidazol-1-ylphenylamine

Hazard: Irritant

Formula: C₉H₉N₃
MW: 159.19 g/mol
MDL Number: MFCD09258808
CAS Number: 112677-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR