2-Hydroxyterephthalic acid dimethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Dimethyl 2-hydroxyterephthalate

059104-1G 059104-5G
101917-100EA 335.38 USD
101917-100 101917-102
2-Hydroxyterephthalic acid dimethyl ester ≥95%
2-Hydroxyterephthalic acid dimethyl ester

Hazard: Irritant

Formula: C₁₀H₁₀O₅
MW: 210.19 g/mol
MDL Number: MFCD08445666
CAS Number: 6342-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR