(R)-2-(2-Hydroxyphenyl)thiazolidine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

059103-1G 059103-5G 059103-10G
101917-094EA 263.08 USD
101917-094 101917-096 101917-098
(R)-2-(2-Hydroxyphenyl)thiazolidine-4-carboxylic acid ≥95%
(R)-2-(2-Hydroxyphenyl)thiazolidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₁NO₃S
MDL Number: MFCD01543538
CAS Number: 201942-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR