N-Hydroxy-2-morpholin-4-yl-2-phenylacetamidine ≥95%

Supplier: Matrix Scientific

059099-1G 059099-5G 059099-10G
101917-082EA 320 USD
101917-082 101917-084 101917-086
N-Hydroxy-2-morpholin-4-yl-2-phenylacetamidine ≥95%
N-Hydroxy-2-morpholin-4-yl-2-phenylacetamidine

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₂
MW: 235.29 g/mol
MDL Number: MFCD11052466
CAS Number: 1053659-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR