6-Hydroxymethylnicotinic acid ethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-(hydroxymethyl)pyridine-3-carboxylate

059096-500MG 059096-1G 059096-5G
101917-066EA 466.15 USD
101917-066 101917-068 101917-070
6-Hydroxymethylnicotinic acid ethyl ester ≥95%
6-Hydroxymethylnicotinic acid ethyl ester

Hazard: Irritant

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
MDL Number: MFCD12131130
CAS Number: 35005-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR