2-Hydroxy-4-methylnicotinic acid ≥95%

Supplier: Matrix Scientific

059095-500MG 059095-1G 059095-5G
101917-060EA 229.23 USD
101917-060 101917-062 101917-064
2-Hydroxy-4-methylnicotinic acid ≥95%
2-Hydroxy-4-methylnicotinic acid

Hazard: Irritant

Formula: C₇H₇NO₃
MDL Number: MFCD09033815
CAS Number: 38076-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR