2-Hydroxy-6-methylnicotinonitrile ≥95%

Supplier: Matrix Scientific

059094-1G 059094-5G
101917-056EA 872.31 USD
101917-056 101917-058
2-Hydroxy-6-methylnicotinonitrile ≥95%
2-Hydroxy-6-methylnicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile

Hazard: Irritant

Formula: C₇H₆N₂O
MW: 134.14 g/mol
MDL Number: MFCD00006013
CAS Number: 4241-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR