5-Hydroxy-1-methyl-1H-pyrazole-3-carboxylic acid methyl ester ≥95%

Supplier: Matrix Scientific

059093-1G 059093-5G 059093-10G
101917-050EA 169.23 USD
101917-050 101917-052 101917-054
5-Hydroxy-1-methyl-1H-pyrazole-3-carboxylic acid methyl ester ≥95%
5-Hydroxy-1-methyl-1H-pyrazole-3-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₆H₈N₂O₃
MDL Number: MFCD01851055
CAS Number: 51985-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR