3-Hydroxy-1-methyl-1H-pyrazole-4-carboxylic acid ethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 1-methyl-3-oxo-2,3-dihydro-1H-pyrazole-4-carboxylate

059092-1G 059092-5G
101917-046EA 521.54 USD
101917-046 101917-048
3-Hydroxy-1-methyl-1H-pyrazole-4-carboxylic acid ethyl ester ≥95%
3-Hydroxy-1-methyl-1H-pyrazole-4-carboxylic acid ethyl ester

Hazard: Irritant

Formula: C₇H₁₀N₂O₃
MW: 170.17 g/mol
Melting Pt: 144…146 °C
MDL Number: MFCD12131137
CAS Number: 103626-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR