2-Hydroxy-5-iodonicotinic acid methyl ester ≥95%

Supplier: Matrix Scientific

059089-500MG 059089-1G 059089-5G
101917-034EA 466.15 USD
101917-034 101917-036 101917-038
2-Hydroxy-5-iodonicotinic acid methyl ester ≥95%
2-Hydroxy-5-iodonicotinic acid methyl ester

Hazard: Irritant

Formula: C₇H₆INO₃
MDL Number: MFCD08235111
CAS Number: 116387-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR