3-Hydroxy-1H-pyrazole-4-carboxylic acid ethyl ester ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 3-hydroxy-1H-pyrazole-4-carboxylate
059088-1G
059088-5G
101917-030EA
144.62
USD
101917-030
101917-032
3-Hydroxy-1H-pyrazole-4-carboxylic acid ethyl ester ≥95%
3-Hydroxy-1H-pyrazole-4-carboxylic acid ethyl ester
Hazard: Irritant
Formula:
C₆H₈N₂O₃ MW: 156.14 g/mol |
MDL Number:
MFCD12131138 CAS Number: 7251-53-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...