2-Hydroxy-4,6-dimethylnicotinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4,6-Dimethyl-2-hydroxynicotinic acid, 4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carboxylic acid

059086-1G 059086-5G
101917-020EA 238.46 USD
101917-020 101917-022
2-Hydroxy-4,6-dimethylnicotinic acid ≥95%
2-Hydroxy-4,6-dimethylnicotinic acid

Hazard: Irritant

Formula: C₈H₉NO₃
MW: 167.17 g/mol
MDL Number: MFCD00234425
CAS Number: 24667-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR