2-Hydroxy-4,6-dimethylnicotinamide ≥95%

Supplier: Matrix Scientific

059085-1G 059085-5G 059085-10G
101917-014EA 263.08 USD
101917-014 101917-016 101917-018
2-Hydroxy-4,6-dimethylnicotinamide ≥95%
2-Hydroxy-4,6-dimethylnicotinamide

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD10568287
CAS Number: 39088-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR