tert-Butyl (2-hydrazinyl-2-oxoethyl)carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Aminoacetohydrazide, 2-BOC protected

059083-1G 059083-5G 059083-10G
101917-004EA 200 USD
101917-004 101917-006 101917-008
tert-Butyl (2-hydrazinyl-2-oxoethyl)carbamate ≥95%
tert-Butyl (2-hydrazinyl-2-oxoethyl)carbamate

Hazard: Irritant

Formula: C₇H₁₅N₃O₃
MW: 189.2100 g/mol
Melting Pt: 122 °C
MDL Number: MFCD01570558
CAS Number: 6926-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR