6H-Furo[2,3-b]pyrrole-5-carboxylic acid methyl ester ≥95%

Supplier: Matrix Scientific

059082-500MG 059082-1G
101917-000EA 253.85 USD
101917-000 101917-002
6H-Furo[2,3-b]pyrrole-5-carboxylic acid methyl ester ≥95%
6H-Furo[2,3-b]pyrrole-5-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₈H₇NO₃
MW: 165.15 g/mol
MDL Number: MFCD11052464
CAS Number: 201019-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR