4H-Furo[3,2-b]pyrrole-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

059081-500MG 059081-1G
101916-996EA 553.85 USD
101916-996 101916-998
4H-Furo[3,2-b]pyrrole-5-carboxylic acid ≥95%
4H-Furo[3,2-b]pyrrole-5-carboxylic acid

Hazard: Irritant

Formula: C₇H₅NO₃
MDL Number: MFCD07368660
CAS Number: 67268-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR