4H-Furo[3,2-b]pyrrole-5-carboxylic acid methylester ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4H-furo[3,2-b]pyrrole-5-carboxylate

059080-1G 059080-5G 059080-10G
101916-990EA 207.69 USD
101916-990 101916-992 101916-994
4H-Furo[3,2-b]pyrrole-5-carboxylic acid methylester ≥95%
4H-Furo[3,2-b]pyrrole-5-carboxylic acid methylester

Hazard: Irritant

Formula: C₈H₇NO₃
MW: 165.15 g/mol
MDL Number: MFCD06797487
CAS Number: 77484-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR