2-(4-Fluorophenoxy)benzonitrile ≥95%

Supplier: Matrix Scientific

059071-500MG 059071-1G
101916-958EA 553.85 USD
101916-958 101916-960
2-(4-Fluorophenoxy)benzonitrile ≥95%
2-(4-Fluorophenoxy)benzonitrile

Hazard: Irritant

Formula: C₁₃H₈FNO
MDL Number: MFCD11155956
CAS Number: 1020922-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR