3-Fluoro-4-methylthiobenzamide ≥95%
Supplier: Matrix Scientific
059069-1G
059069-5G
101916-952EA
106.15
USD
101916-952
101916-954
3-Fluoro-4-methylthiobenzamide ≥95%
3-Fluoro-4-methylthiobenzamide
Hazard: Irritant
Formula:
C₈H₈FNS MW: 169.22 g/mol |
MDL Number:
MFCD00275549 CAS Number: 175277-87-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...