2-Fluoroisonicotinoyl chloride ≥95%

Supplier: Matrix Scientific

059068-1G 059068-5G
101916-948EA 453.85 USD
101916-948 101916-950
2-Fluoroisonicotinoyl chloride ≥95%
2-Fluoroisonicotinoyl chloride

Hazard: Irritant

Formula: C₆H₃ClFNO
MDL Number: MFCD11227178
CAS Number: 65352-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR