6-Fluoro-8-isocyanato-4H-benzo[1,3]dioxine ≥95%

Supplier: Matrix Scientific

059067-1G 059067-5G 059067-10G
101916-940EA 183.08 USD
101916-940 101916-942 101916-944
6-Fluoro-8-isocyanato-4H-benzo[1,3]dioxine ≥95%
6-Fluoro-8-isocyanato-4H-benzo[1,3]dioxine

Hazard: Irritant

Formula: C₉H₆FNO₃
MDL Number: MFCD02681891
CAS Number: 321309-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR