6-Fluoro-4H-benzo[1,3]dioxine-8-carboxylic acid ≥95%

Supplier: Matrix Scientific

059066-1G 059066-10G
101916-936EA 320 USD
101916-936 101916-938
6-Fluoro-4H-benzo[1,3]dioxine-8-carboxylic acid ≥95%
6-Fluoro-4H-benzo[1,3]dioxine-8-carboxylic acid

Hazard: Irritant

Formula: C₉H₇FO₄
MW: 198.15 g/mol
MDL Number: MFCD02681981
CAS Number: 321309-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR