Ethyl-5-(2-tert-butyloxycarbonylaminoethyl)[1,2,4]oxadiazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

059058-500MG 059058-1G 059058-5G
101916-902EA 466.15 USD
101916-902 101916-904 101916-906
Ethyl-5-(2-tert-butyloxycarbonylaminoethyl)[1,2,4]oxadiazole-3-carboxylate ≥95%
Ethyl-5-(2-tert-butyloxycarbonylaminoethyl)[1,2,4]oxadiazole-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₉N₃O₅
MW: 285.3 g/mol
MDL Number: MFCD10568205
CAS Number: 652158-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR