Ethyl-5-(tert-butyloxycarbonylaminomethyl)[1,3,4]oxadiazole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-(((tert-butoxycarbonyl)amino)methyl)-1,3,4-oxadiazole-2-carboxylate

059057-1G 059057-5G
101916-898EA 356.92 USD
101916-898 101916-900
Ethyl-5-(tert-butyloxycarbonylaminomethyl)[1,3,4]oxadiazole-2-carboxylate ≥95%
Ethyl-5-(tert-butyloxycarbonylaminomethyl)[1,3,4]oxadiazole-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₇N₃O₅
MW: 271.28 g/mol
MDL Number: MFCD10568221
CAS Number: 164029-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR