Ethyl-1-phenyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 1-phenyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate
059054-1G
059054-5G
059054-10G
101916-882EA
263.08
USD
101916-882
101916-884
101916-886
Ethyl-1-phenyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate ≥95%
Ethyl-1-phenyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate
Hazard: Irritant
Formula:
C₁₃H₁₁F₃N₂O₂ MW: 284.2339 g/mol |
MDL Number:
MFCD10568308 CAS Number: 741717-63-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...