Ethyl-3-phenyl-[1,2,4]oxadiazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-phenyl-1,2,4-oxadiazole-5-carboxylate

059053-1G 059053-5G 059053-10G
101916-876EA 369.23 USD
101916-876 101916-878 101916-880
Ethyl-3-phenyl-[1,2,4]oxadiazole-5-carboxylate ≥95%
Ethyl-3-phenyl-[1,2,4]oxadiazole-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₃
MW: 218.21 g/mol
MDL Number: MFCD03232737
CAS Number: 37760-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR