Ethyl-2-(4-methoxyphenoxy)acetoacetate ≥95%

Supplier: Matrix Scientific

059049-1G 059049-5G
101916-860EA 436.92 USD
101916-860 101916-862
Ethyl-2-(4-methoxyphenoxy)acetoacetate ≥95%
Ethyl-2-(4-methoxyphenoxy)acetoacetate

Hazard: Irritant

Formula: C₁₃H₁₆O₅
MDL Number: MFCD10568346
CAS Number: 7699-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR