Ethylsulfamoyl chloride ≥95%
Supplier: Matrix Scientific
059048-500MG
059048-1G
101916-856EA
436.92
USD
101916-856
101916-858
Ethylsulfamoyl chloride ≥95%
Ethylsulfamoyl chloride
Hazard: Irritant/Refrigerate
Formula:
C₂H₆ClNO₂S Boiling Pt: 96…98 °C (0.2 Torr) |
MDL Number:
MFCD12401754 CAS Number: 16548-07-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...