Ethyl-5-hydroxymethyl-3-methylisoxazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

059047-1G 059047-5G
101916-852EA 553.85 USD
101916-852 101916-854
Ethyl-5-hydroxymethyl-3-methylisoxazole-4-carboxylate ≥95%
Ethyl-5-hydroxymethyl-3-methylisoxazole-4-carboxylate

Hazard: Irritant

Formula: C₈H₁₁NO₄
MW: 185.18 g/mol
Boiling Pt: 125…130 °C (1 Torr)
Melting Pt: 35…36 °C
MDL Number: MFCD10568348
CAS Number: 95104-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR