Ethyl-5-formyl-3-methylisoxazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

059046-1G 059046-5G
101916-848EA 378.46 USD
101916-848 101916-850
Ethyl-5-formyl-3-methylisoxazole-4-carboxylate ≥95%
Ethyl-5-formyl-3-methylisoxazole-4-carboxylate

Hazard: Irritant

Formula: C₈H₉NO₄
MW: 183.17 g/mol
MDL Number: MFCD10568350
CAS Number: 129663-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR