Ethyl-5-formylisoxazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

059045-1G 059045-5G
101916-844EA 421.54 USD
101916-844 101916-846
Ethyl-5-formylisoxazole-3-carboxylate ≥95%
Ethyl-5-formylisoxazole-3-carboxylate

Hazard: Irritant

Formula: C₇H₇NO₄
MW: 169.14 g/mol
Melting Pt: 35…37 °C
MDL Number: MFCD11052378
CAS Number: 22667-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR