Ethyl-1-ethoxycarbonylmethyl-iso-nipecotate ≥95%

Supplier: Matrix Scientific

059043-1G 059043-5G 059043-10G
101916-832EA 378.46 USD
101916-832 101916-834 101916-836
Ethyl-1-ethoxycarbonylmethyl-iso-nipecotate ≥95%
Ethyl-1-ethoxycarbonylmethyl-iso-nipecotate

Hazard: Irritant

Formula: C₁₂H₂₁NO₄
MDL Number: MFCD00101929
CAS Number: 1838-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR