Ethyl acetopyruvate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2,4-dioxopentanoate, Ethyl 2,4-dioxovalerate

059041-25G 059041-100G
101916-826EA 96.92 USD
101916-826 101916-828
Ethyl acetopyruvate ≥95%
Ethyl acetopyruvate

Hazard: Irritant

Formula: C₇H₁₀O₄
MW: 158.15 g/mol
Boiling Pt: 108-109°C/12mm
Melting Pt: 16-18°C
Density: 1.126
MDL Number: MFCD00009124
CAS Number: 615-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR