Ethyl-5-cyano-6-hydroxy-2-methylnicotinate ≥95%

Supplier: Matrix Scientific

059039-1G 059039-5G 059039-10G
101916-816EA 378.46 USD
101916-816 101916-818 101916-820
Ethyl-5-cyano-6-hydroxy-2-methylnicotinate ≥95%
Ethyl-5-cyano-6-hydroxy-2-methylnicotinate

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₃
MDL Number: MFCD10568202
CAS Number: 52600-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR