Ethyl-1-(2-cyanoethyl)isonipecotate ≥95%

Supplier: Matrix Scientific

059038-1G 059038-5G
101916-812EA 263.08 USD
101916-812 101916-814
Ethyl-1-(2-cyanoethyl)isonipecotate ≥95%
Ethyl-1-(2-cyanoethyl)isonipecotate

Hazard: Irritant

Formula: C₁₁H₁₈N₂O₂
MDL Number: MFCD10695713
CAS Number: 477787-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR