Ethyl-1-(3-chloro-5-(trifluoromethyl)pyridin-2-yl)isonipecotate ≥95%

Supplier: Matrix Scientific

059035-1G 059035-5G
101916-796EA 346.15 USD
101916-796 101916-798
Ethyl-1-(3-chloro-5-(trifluoromethyl)pyridin-2-yl)isonipecotate ≥95%
Ethyl-1-(3-chloro-5-(trifluoromethyl)pyridin-2-yl)isonipecotate

Hazard: Irritant

Formula: C₁₄H₁₆ClF₃N₂O₂
MDL Number: MFCD06409177
CAS Number: 874814-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR